Learn More
JARID1A/RBP2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | JARID1A/RBP2 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2164
![]() |
Bethyl Laboratories, Inc
IHC00479T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2163
![]() |
Bethyl Laboratories, Inc
IHC00479 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Dog, Horse, Rabbit, Guinea pig_10141, Panda, Orangutan, Gorilla and Northern w
This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.Specifications
JARID1A/RBP2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
histone demethylase JARID1A, JARID1A, jumonji, AT rich interactive domain 1A, Jumonji, AT rich interactive domain 1A (RBBP2-like), Jumonji, AT rich interactive domain 1A (RBP2-like), jumonji/ARID domain-containing protein 1A, lysine (K)-specific demethylase 5A, RBBP2, RBBP-2, RBP2, retinoblastoma binding protein 2, retinoblastoma-binding protein 2 | |
KDM5A | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P29375 | |
5927 | |
Between 1640 and 1690 | |
Primary | |
4° C | |
KDM5A |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.