Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JARID1C Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140593
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9074 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9074 Supplier Bethyl Laboratories, Inc Supplier No. A301035AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Dog and Pig

This gene is a member of the SMCY homolog family and encodes a protein with one ARID domain, one JmjC domain, one JmjN domain and two PHD-type zinc fingers. The DNA-binding motifs suggest this protein is involved in the regulation of transcription and chromatin remodeling. Mutations in this gene have been associated with X-linked mental retardation. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.

Specifications

Antigen JARID1C
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene KDM5C
Gene Accession No. P41229
Gene Alias DXS1272E, JARID1C, MRXJ, MRXSCJ, MRXSJ, SMCX, XE169, JmjC domain-containing protein SMCX, Jumonji, AT rich interactive domain 1C (RBP2-like), Jumonji/ARID domain-containing protein 1C, Smcx homolog, X chromosome, Smcy homolog, X-linked, histone demethylase JARID1C, lysine-specific demethylase 5C, protein SmcX, sele
Gene Symbols KDM5C
Host Species Rabbit
Immunogen Between 1510 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8242
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.