Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Jasplakinolide
GSA_VAGREENER_CHOICESDP

Catalog No. 2792100U Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-92100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-92100U Supplier Tocris Bioscience™ Supplier No. 2792/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Stabilizes F-actin; promotes actin polymerization

Rapidly stabilizes pre-formed actin filaments and inhibits their disassembly in vitro. Also induces polymerization of actin monomers into F-actin in vivo. Shown to bind to F-actin competitively with phalloidin (Cat. No. 4535) (Kd ∽ 15 nM). Exhibits antifungal and antiproliferative effects (IC50 = 35 nM for antiproliferative activity in PC3 cells). Cell permeable.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 102396-24-7
Molecular Formula C36H45BrN4O6

Specifications

Quantity 100 μg
Formula Weight 709.67
Purity >97%
Molecular Weight (g/mol) 709.682
Chemical Name or Material Jasplakinolide
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.