Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JAZF1 Antibody (PCRP-JAZF1-1C2), FITC, Novus Biologicals™
SDP

Catalog No. NB157006 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB157006 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB157006 Supplier Novus Biologicals Supplier No. NBP314221F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

JAZF1 Monoclonal antibody specifically detects JAZF1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen JAZF1
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-JAZF1-1C2
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias JAZF zinc finger 1, juxtaposed with another zinc finger gene 1, juxtaposed with another zinc finger protein 1, TAK1-interacting protein 27, TIP27ZNF802DKFZp761K2222, Zinc finger protein 802
Host Species Mouse
Immunogen Recombinant fragment (around aa1-130) of human JAZF1 protein (exact sequence is proprietary) (Uniprot: Q86VZ6)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 221895
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.