Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Jedi2

Click to view available options
Quantity:
10 mg
50 mg
Description
Jedi2 is a piezo1 channel activator. Promotes Ca2+ influx into HEK293 cells expressing Piezo1 channels. Elicits responses in mouse Piezo1-transfected, but not Piezo2-transfected cells.
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-5-(2-thienyl)-3-furancarboxylic acid |
| CAS | 651005-90-2 |
| Quantity | 10 mg |
| Molecular Formula | C10H8O3S |
| Formula Weight | 208.23 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction