Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ JH-II-127

missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
JH-II-127 is a potent and selective LRRK2 inhibitor (IC50 values are 2.2, 6.6 and 47.7 nM for G2019S-mutant, wild-type and A2016T-mutant LRRK2, respectively). Selective for LRRK2 over a panel of 138 kinases. Brain penetrant and orally bioavailable.
Specifications
Specifications
Chemical Name or Material | [4-[[5-Chloro-4-(methylamino)-7H-pyrrolo[2,3-d]pyrimidin-2-yl]amino]-3-methoxyphenyl]-4-morpholinyl-methanone |
CAS | 1700693-08-8 |
Assay Percent Range | >98% |
Quantity | 10 mg |
Molecular Formula | C19H21ClN6O3 |
Formula Weight | Observed MW: 416.87 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction