Learn More
Description
Specifications
Specifications
| Antigen | JHDM1D |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | jmjC domain-containing histone demethylation protein 1D, jumonji C domain containing histone demethylase 1 homolog D (S. cerevisiae), KDM7A |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 417-735 of mouse KDM7A JHDM1D (NP_001028602.2). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
