Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ JIB 04

Click to view available options
Quantity:
10 mg
50 mg
Description
JIB 04 is a pan Jumonji histone demethylase inhibitor (IC50 values are 230, 340, 435, 445, 855 and 1100 nM for JARID1A, JMJD2E, JMJD2B, JMJD2A, JMJD3 and JMJD2C respectively). Exhibits negligible activity at PHD2 and TET1. Selectively blocks cancer cell growth in vitro and diminishes tumor growth in H358 and A549 mouse xenograft models in vivo. Prolongs survival in a mouse model of breast cancer.
Specifications
Specifications
Quantity | 50 mg |
Product Type | JIB 04 |
Purity | >99% |
Molecular Weight (g/mol) | 308.76 |
Inhibitors | JIB 04 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction