Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD2D Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP103357C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP103357C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP103357C Supplier Novus Biologicals Supplier No. NBP103357C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JMJD2D Polyclonal specifically detects JMJD2D in Human samples. It is validated for Western Blot, ICC/IF (Negative).

Specifications

Antigen JMJD2D
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, ICC/IF (Negative)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 1.14.11, EC 1.14.11.-, FLJ10251, JHDM3D, JmjC domain-containing histone demethylation protein 3D, JMJD2Dlysine-specific demethylase 4D, jumonji domain containing 2D, Jumonji domain-containing protein 2D, lysine (K)-specific demethylase 4D, MGC141909
Gene Symbols KDM4D
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human JMJD2D protein (within residues 460-520). [Swiss-Prot# Q6B0I6]
Molecular Weight of Antigen 55 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 55693
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.