Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD2D Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP24941125UL
Description
JMJD2D Polyclonal antibody specifically detects JMJD2D in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
JMJD2D | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
EC 1.14.11, EC 1.14.11.-, FLJ10251, JHDM3D, JmjC domain-containing histone demethylation protein 3D, JMJD2Dlysine-specific demethylase 4D, jumonji domain containing 2D, Jumonji domain-containing protein 2D, lysine (K)-specific demethylase 4D, MGC141909 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PLLAGTTCTASGPEPEPLPEDGALMDKPVPLSPGLQHPVKASGCSWAPV | |
25 μL | |
Transcription Factors and Regulators | |
55693 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction