Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JMJD2D Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $620.58
Specifications
Antigen | JMJD2D |
---|---|
Concentration | Lot-Specific |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2654
![]() ![]() |
Active Motif
39248 |
10 μL |
Each for $156.65
|
|
|||||
50-199-2672
![]() ![]() |
Active Motif
39247 |
100 μL |
Each for $620.58
|
|
|||||
Description
JMJD2D belongs to JMJD2 family that is consist of four cancer-associated genes.Specifications
JMJD2D | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6B0I6 | |
55693 | |
This JMJD2D antibody was raised against a peptide corresponding to amino acids 337-359 of human JMJD2D. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
KDM4D |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
FLJ10251; Jhdm3d; JmjC domain-containing histone demethylation protein 3D; Jmjd2d; jumonji domain containing 2D; Jumonji domain-containing protein 2D; Kdm4d; lysine (K)-specific demethylase 4D; lysine demethylase 4D; Lysine-specific demethylase 4D | |
KDM4D | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title