Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMJD4 Antibody (JMJD4/32b/H12), Biotin, Novus Biologicals™
SDP

Catalog No. NBP250425B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250425B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250425B Supplier Novus Biologicals Supplier No. NBP250425B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JMJD4 Monoclonal specifically detects JMJD4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Knockdown Validated.

Specifications

Antigen JMJD4
Applications Western Blot, ELISA, Immunohistochemistry, KnockDown
Classification Monoclonal
Clone JMJD4/32b/H12
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ12517, jumonji domain containing 4, jumonji domain-containing protein 4
Gene Symbols JMJD4
Host Species Mouse
Immunogen Recombinant protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 65094
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.