Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JMY Antibody (HMY117), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP250457IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250457IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250457IR Supplier Novus Biologicals Supplier No. NBP250457IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JMY Monoclonal specifically detects JMY in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen JMY
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone HMY117
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ37870, junction mediating and regulatory protein, p53 cofactor, junction-mediating and regulatory protein, junction-mediating and -regulatory protein, MGC163496, WAS protein homology region 2 domain containing 1-like 3, WHDC1L3
Gene Symbols JMY
Host Species Mouse
Immunogen Synthetic peptide from the c terminus of the peptide sequence, identical for both isoform 1 and 2
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 133746
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.