Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JNJ 303
GREENER_CHOICE

Catalog No. 389910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-991-0 10 mg
38-995-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-991-0 Supplier Tocris Bioscience™ Supplier No. 3899/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective IKs blocker

Potent IKs blocker (IC50 = 64 nM). Displays no effects on other cardiac channels; IC50 values are 3.3, >10, 11.1 and 12.6 μM for INa, ICa, Ito and IKr currents respectively. Prolongs QT and causes unprovoked torsades de pointes (TdP).

Chemical Identifiers

CAS 878489-28-2
Molecular Formula C21H29ClN2O4S

Specifications

Quantity 10 mg
Formula Weight 440.98
Purity >98%
Molecular Weight (g/mol) 440.983
Chemical Name or Material JNJ 303
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.