Learn More
Description
Specifications
Specifications
| Antigen | JNK1 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500 |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Alias | c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8JNK, protein kinase JNK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 |
| Gene Symbols | MAPK8 |
| Host Species | Rabbit |
| Immunogen | A recombinant protein made to the C-terminus (residues 228-451) of the C. elegans JNK1 alpha protein. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
