Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNK1 Antibody (OTI4E3), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP271256IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271256IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271256IR Supplier Novus Biologicals Supplier No. NBP271256IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JNK1 Monoclonal specifically detects JNK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen JNK1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4E3
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8JNK, protein kinase JNK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1
Gene Symbols MAPK8
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-384 of human JNK1 (NP_002741) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Hypoxia, Immunology, Innate Immunity, MAP Kinase Signaling, p53 Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5599.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.