Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JNK1, p Thr183 Antibody, Novus Biologicals™
SDP

Catalog No. NB017857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017857 0.025 mL
NB10082009 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017857 Supplier Novus Biologicals Supplier No. NB100820090.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

JNK1 (p Thr183) Polyclonal specifically detects JNK1 (p Thr183) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen JNK1 (p Thr183)
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Formulation PBS [pH 7.4] 150mM NaCl and 50% glycerol (without Mg2+ and Ca2+) with 0.02% Sodium Azide
Gene Accession No. P45984/P53779
Gene Alias c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8JNK, protein kinase JNK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1
Gene Symbols MAPK8
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human JNK2/3/SAPK around the phosphorylation site of threonine 183 (M-M-TP-P-Y).
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Hypoxia, MAP Kinase Signaling, p53 Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5599
Test Specificity SAPK/JNK (specific to Phospho-Thr183) detects endogenous levels of SAPK/JNK only when phosphorylated at threonine 183.
Target Species Human, Rat, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.