Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JNK3 Antibody (2B10H12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261801
Description
JNK3 Monoclonal specifically detects JNK3 in Human, Rat samples. It is validated for Western Blot, ELISA.Specifications
JNK3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
c-Jun N-terminal kinase 3, EC 2.7.11, EC 2.7.11.24, JNK3 alpha protein kinase, JNK3A, JNK3FLJ12099, MAP kinase 10, MAP kinase p49 3F12, MAPK 10, MGC50974, mitogen-activated protein kinase 10, p493F12, p54bSAPK, PRKM10FLJ33785, stress activated protein kinase beta, Stress-activated protein kinase JNK3 | |
Mouse | |
52.6 kDa | |
0.1 mL | |
Primary | |
Human, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
2B10H12 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
MAPK10 | |
Purified recombinant fragment of human MAPK10 (AA: 180-329) expressed in E. Coli. | |
Protein G purified | |
Immunology, Innate Immunity, Protein Kinase, Wnt Signaling Pathway | |
5602 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction