Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JNK3 Antibody (2B10H12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | JNK3 |
---|---|
Clone | 2B10H12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61801
![]() |
Novus Biologicals
NBP261801 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
JNK3 Monoclonal specifically detects JNK3 in Human, Rat samples. It is validated for Western Blot, ELISA.Specifications
JNK3 | |
1 mg/mL | |
Monoclonal | |
Purified | |
Immunology, Innate Immunity, Protein Kinase, Wnt Signaling Pathway | |
c-Jun N-terminal kinase 3, EC 2.7.11, EC 2.7.11.24, JNK3 alpha protein kinase, JNK3A, JNK3FLJ12099, MAP kinase 10, MAP kinase p49 3F12, MAPK 10, MGC50974, mitogen-activated protein kinase 10, p493F12, p54bSAPK, PRKM10FLJ33785, stress activated protein kinase beta, Stress-activated protein kinase JNK3 | |
MAPK10 | |
IgG2a | |
Protein G purified |
2B10H12 | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
Unconjugated | |
Mouse | |
Human, Rat | |
5602 | |
Purified recombinant fragment of human MAPK10 (AA: 180-329) expressed in E. Coli. | |
Primary | |
52.6 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title