Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ (+)-JQ1 PA
Click to view available options
Quantity:
25 mg
Description
(+)-JQ1 PA is a click-activated (alkyne) version of the BET bromodomain inhibitor (+)-JQ1 (Cat. No. 4499). Can be used as a precursor to a PROTAC® targeting BET bromodomains. Supplied with an alkyne functionalized handle for click conjugation to a linker/E3 ligase ligand. Can also be conjugated to fluorescent dyes for imaging applications.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
Chemical Name or Material | (6S)-4-(4-Chlorophenyl)-2,3,9-trimethyl-N-2-propyn-1-yl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetamide |
CAS | 2115701-93-2 |
Quantity | 25 mg |
Molecular Formula | C22H20ClN5OS |
Formula Weight | 437.95 |
Purity | 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction