Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ JQEZ5

Click to view available options
Quantity:
10 mg
50 mg
Description
JQEZ5 is a potent and selective SAM-competitive EZH2 lysine methyltransferase inhibitor (IC50 = 11 nM). Selectively binds EZH2 over a panel of 22 methyltransferases. Reduces global level of the PRC2-associated mark H3K27me3 in K562 cells. Suppresses cell growth in CML cells. Inhibits colony formation of primary human CD34+ CML stem/ progenitor cells. Antitumor effects in vivo.
Specifications
Specifications
| Chemical Name or Material | N-[(1,2-Dihydro-6-methyl-2-oxo-4-propyl-3-pyridinyl)methyl]-1-(1-methylethyl)-6-[6-(4-methyl-1-piperazinyl)-3-pyridinyl]-1H-pyrazolo[3,4-b]pyridine-4-carboxamide |
| CAS | 1913252-04-6 |
| Quantity | 10 mg |
| Molecular Formula | C30H38N8O2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction