Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ JTV 519 fumarate

Click to view available options
Quantity:
10 mg
50 mg
Description
JTV 519 fumarate is a ryanodine receptor (RyR) inhibitor; stabilizes RyR2 in a closed conformation. Exhibits antiarrhythmic and cardioprotective properties in vivo.
Specifications
Specifications
Quantity | 10 mg |
Product Type | JTV 519 fumarate |
Purity | 98% |
Molecular Weight (g/mol) | M.W. (Observed): 540.67 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction