Learn More
Description
Specifications
Specifications
| Antigen | Jumonji/JARID2 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | JMJjumonji-like protein, jumonji (mouse) homolog, jumonji homolog, jumonji, AT rich interactive domain 2, Jumonji/ARID domain-containing protein 2, protein Jumonji |
| Gene Symbols | JARID2 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to an N-terminal portion of the human JARID2 protein sequence (between residues 1-100). [UniProtNo. Q92833] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
