Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Jumonji/JARID2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB1002214F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002214F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002214F Supplier Novus Biologicals Supplier No. NB1002214F

Rabbit Polyclonal Antibody

Jumonji/JARID2 Polyclonal specifically detects Jumonji/JARID2 in Human, Mouse, Rabbit samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.

Specifications

Antigen Jumonji/JARID2
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q92833
Gene Alias JMJjumonji-like protein, jumonji (mouse) homolog, jumonji homolog, jumonji, AT rich interactive domain 2, Jumonji/ARID domain-containing protein 2, protein Jumonji
Gene Symbols JARID2
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human JARID2 protein sequence (between residues 1-100). [UniProt# Q92833]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, Epigenetics, Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3720
Test Specificity Human and mouse. Rabbit reactivity reported in scientific literature (PMID: 24074864).
Target Species Human, Mouse, Rabbit
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.