Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JunB/AP-1 Antibody (5H4), Novus Biologicals™
SDP

Catalog No. NB298606 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298606 50 μL
NB298607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298606 Supplier Novus Biologicals Supplier No. NBP32618550UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

JunB/AP-1 Monoclonal antibody specifically detects JunB/AP-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen JunB/AP-1
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 5H4
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B
Host Species Rabbit
Immunogen A synthesized peptide derived from Human JunB/AP-1 [UniProt P17275]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3726
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.