Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JunB/AP-1 Antibody (5H4), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | JunB/AP-1 |
---|---|
Clone | 5H4 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Description
JunB/AP-1 Monoclonal antibody specifically detects JunB/AP-1 in Human samples. It is validated for ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
JunB/AP-1 | |
ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B | |
A synthesized peptide derived from Human JunB/AP-1 [UniProt P17275] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5H4 | |
ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3726 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title