Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JunB/AP-1 Antibody (PCRP-JUNB-3G11), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB380893 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380893 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380893 Supplier Novus Biologicals Supplier No. NBP308390AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

JunB/AP-1 Monoclonal antibody specifically detects JunB/AP-1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen JunB/AP-1
Applications Flow Cytometry, ELISA, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-JUNB-3G11
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B
Host Species Mouse
Immunogen Recombinant full-length human JunB/AP-1 protein (Uniprot: P17275)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3726
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.