Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JunB/AP-1 Mouse anti-Human, Clone: PCRP-JUNB-3G2, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30757920UG
Description
JunB/AP-1 Monoclonal specifically detects JunB/AP-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
JunB/AP-1 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B | |
Recombinant full-length human JunB/AP-1 protein | |
20 μg | |
Phospho Specific | |
3726 | |
Store at 4°C. | |
IgG1 |
Flow Cytometry, Immunofluorescence, Peptide Array | |
PCRP-JUNB-3G2 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction