Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JunB/AP-1 Mouse anti-Human, Clone: PCRP-JUNB-3G2, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen JunB/AP-1
Clone PCRP-JUNB-3G2
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Applications Flow Cytometry, Immunofluorescence, Peptide Array
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122632
SDP
View Documents
Novus Biologicals
NBP30757920UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

JunB/AP-1 Monoclonal specifically detects JunB/AP-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications

Specifications

JunB/AP-1
0.2 mg/ml
Flow Cytometry, Immunofluorescence, Peptide Array
Unconjugated
Mouse
Phospho Specific
10 mM PBS with 0.05% BSA
3726
IgG1
Protein A or G purified
PCRP-JUNB-3G2
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
Human
activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B
Recombinant full-length human JunB/AP-1 protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.