Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JunB Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $620.58
Specifications
Antigen | JunB |
---|---|
Concentration | Conc. Not Determined |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-4047
![]() ![]() |
Active Motif
39550 |
10 μL |
Each for $156.65
|
|
|||||
50-199-4046
![]() ![]() |
Active Motif
39549 |
200 μL |
Each for $620.58
|
|
|||||
Description
JunB ,a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5'-TGA[CG]TCA-3'. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.Specifications
JunB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P17275 | |
3726 | |
This JunB antibody was raised against a synthetic peptide corresponding to amino acids 28-42 of human JunB. | |
Primary | |
Antibody |
Conc. Not Determined | |
Polyclonal | |
Liquid | |
RUO | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
activator protein 1; AP-1; Jun B; jun B proto-oncogene; JUNB; Jun-b; Jun-B oncogene; JunB proto-oncogene, AP-1 transcription factor subunit; MyD21; Oncogene Jun-B; Transcription factor jun-B | |
Junb | |
IgG | |
-20°C, Avoid Freeze/Thaw Cycles | |
Junb |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title