Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JunD Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222439 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
50 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222439 200 μg
NB437690 50 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222439 Supplier Novus Biologicals Supplier No. NBP222439
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JunD Polyclonal specifically detects JunD in Mouse samples. It is validated for ChIP Assay, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen JunD
Applications ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 10 ug/ml, Flow Cytometry 3 - 5 ug/ 10^6 cells, Immunocytochemistry/Immunofluorescence 2 - 3 ug/ml, Chromatin Immunoprecipitation (ChIP) 1-3 ul
Formulation PBS with 0.05% Sodium Azide
Gene Alias activator protein 1, AP-1, jun D proto-oncogene, JunD-FL isoform, transcription factor jun-D
Gene Symbols JUND
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues G(329) C Q L L P Q H Q V P A Y(341) of mouse JunD.
Purification Method Immunogen affinity purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3727
Reconstitution Reconstitute with 0.2 ml PBS to a final concentration of 1 mg/ml.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.