Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JunD Rabbit anti-Human, Clone: SD0830, Novus Biologicals™
SDP

Catalog No. NBP267826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267826 Supplier Novus Biologicals Supplier No. NBP267826

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

JunD Monoclonal specifically detects JunD in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen JunD
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SD0830
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias activator protein 1, AP-1, jun D proto-oncogene, JunD-FL isoform, transcription factor jun-D
Gene Symbols JUND
Host Species Rabbit
Immunogen Recombinant protein within Human JunD aa 101-200/ 347. (SwissProt: P17535 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3727
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.