Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Jurkat Whole Cell Lysate

Description
The Jurkat Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
Tissue | Jurkat |
For Use With (Application) | Western Blot |
Storage Buffer | Supplied in SDS sample buffer containing 5% Beta-mercaptoethanol. |
Test Specificity | Jurkat, clone E6-1 (Human Acute T Cell Leukemia) |
Host Species | Human |
Concentration | 2 mg/mL |
Content And Storage | Store at −80°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction