Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT3 Antibody (S180-41), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB259319647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259319647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259319647 Supplier Novus Biologicals Supplier No. NBP259319AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EAAT3 Monoclonal antibody specifically detects EAAT3 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen EAAT3
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S180-41
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1
Gene Symbols SLC1A1
Host Species Mouse
Immunogen Fusion protein amino acids 1-524 (full length) of human EAAT3 (Uniprot: P43005)
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 6505
Test Specificity Detects 60kDa.
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.