Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JWA Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen JWA
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB124893
SDP
View Documents
Novus Biologicals
NBP309996100UL
100 μg
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

JWA Polyclonal specifically detects JWA in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

JWA
Western Blot
Unconjugated
Rabbit
Mouse
ADP-ribosylation factor-like protein 6-interacting protein 5, ADP-ribosylation-like factor 6 interacting protein 5, Aip-5, ARL-6-interacting protein 5, Cytoskeleton-related vitamin A-responsive protein, Dermal papilla-derived protein 11, DERP11addicsin, Glutamate transporter EAAC1-interacting protein, glutamate transporter EEAC1-associated protein, GTRAP3-18aip-5, hp22, HSPC127, JM5, jmx, JWAcytoskeleton related vitamin A responsive protein, PRA1 domain family 3, PRA1 family protein 3, PRA2, PRAF3dermal papilla derived protein 11, Prenylated Rab acceptor protein 2, Protein JWa, putative MAPK activating protein PM27, Putative MAPK-activating protein PM27
The immunogen is a synthetic peptide directed towards the N terminal region of human JWA (NP_075368.1). Peptide sequence MDVNLAPLRAWDDFFPGSDRFARPDFRDISKWNNRVVSNLLYYQTNYLVV
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
10550
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.