Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ K 02288
GREENER_CHOICE

Catalog No. p-5545842
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Type 1 BMP receptor inhibitor

K 02288 is a potent and selective inhibitor of type I bone morphogenic protein (BMP) receptors (IC50 values are 1.1, 1.8, 6.4, 34.4, 220, 302 and 321 nM for ALK2, ALK1, ALK6, ALK3, ActRIIA, ALK4 and ALK5 respectively). Reduces BMP4-induced Smad1/5/8 phosphorylation in vitro (IC50 = 100 nM) and induces dorsalization of zebrafish embryos.

Specifications

Quantity 10 mg
Product Type K 02288
Purity >99%
Molecular Weight (g/mol) 352.38
Inhibitors K 02288

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.