Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ K 114
GREENER_CHOICESDP

Catalog No. 314450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-445-0 50 mg
31-441-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 31-445-0 Supplier Tocris Bioscience™ Supplier No. 3144/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Amyloid fibril-specific fluorescent dye

Potent amyloid fibril-specific fluorescent dye (EC50 = 20 - 30 nM). Exhibits minimal fluorescence in aqueous buffers and fluoresces brightly in the presence of Aβ, α-synuclein and tau in situ. (Optimum wavelength = 550 nm).

Chemical Identifiers

CAS 872201-12-2
Molecular Formula C22H17BrO2

Specifications

Quantity 50 mg
Formula Weight 393.27
Purity >98%
Molecular Weight (g/mol) 393.28
Chemical Name or Material K 114
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.