Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ K 252a
GREENER_CHOICE

Catalog No. 1683200u
Click to view available options
Quantity:
200 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Protein kinase inhibitor

Non-selective protein kinase inhibitor; analog of staurosporine (Cat.No. 1285). Inhibits PKC (IC50 = 32.9 nM), Ca2+/calmodulin-stimulated phosphodiesterases (IC50 = 1.3 - 2.9 μM), MLCK (Ki = 20 nM) and receptor tyrosine kinases. Inhibits the oncogenic properties of MET; prevents autophosphorylation and activation of downstream effectors (MAPK, Akt).

Chemical Identifiers

CAS 99533-80-9
Molecular Formula C27H21N3O5

Specifications

Quantity 200 μg
Formula Weight 467.48
Purity >98%
Molecular Weight (g/mol) 467.481
Chemical Name or Material K 252a
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.