Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K15 (a putative transmembrane protein similar to LMP2A) Antibody (IF3/A10), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP20341A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20341A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20341A94 Supplier Novus Biologicals Supplier No. NBP250341AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

K15 (a putative transmembrane protein similar to LMP2A) Monoclonal specifically detects K15 (a putative transmembrane protein similar to LMP2A) in Human, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen K15 (a putative transmembrane protein similar to LMP2A)
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone IF3/A10
Conjugate Alexa Fluor 594
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Recombinant protein
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Apoptosis, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Human, Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.