Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KA1/GRIK4/Glutamate Receptor KA1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB357224 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357224 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357224 Supplier Novus Biologicals Supplier No. NBP176851MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KA1/GRIK4/Glutamate Receptor KA1 Polyclonal antibody specifically detects KA1/GRIK4/Glutamate Receptor KA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen KA1/GRIK4/Glutamate Receptor KA1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EAA1, Excitatory amino acid receptor 1, glutamate receptor KA1, Glutamate receptor KA-1, glutamate receptor, ionotropic kainate 4, glutamate receptor, ionotropic, kainate 4, KA1GRIK
Host Species Rabbit
Immunogen Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of the mouse Grik4. The immunogen is located within the last 50 amino acids of Grik4. Amino Acid Squence: RPSPARSEESLEWDKTTN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2900
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.