Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KA2/GRIK5/Glutamate Receptor KA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP26897525UL
Description
KA2/GRIK5/Glutamate Receptor KA2 Polyclonal antibody specifically detects KA2/GRIK5/Glutamate Receptor KA2 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
KA2/GRIK5/Glutamate Receptor KA2 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
EAA2, Excitatory amino acid receptor 2, glutamate receptor KA2, Glutamate receptor KA-2, glutamate receptor, ionotropic kainate 5, glutamate receptor, ionotropic, kainate 5, KA2GRIK2 | |
This antibody was developed against a recombinant protein corresponding to amino acids: KVSTIIIDANASISHLILRKASELGMTSAFYKYILTTMDFPILHLDGIVEDSSNILGFSMFNTSHPFYPEFVRSLNMSWRENCEASTY | |
25 μL | |
Neuroscience | |
2901 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction