Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kaiso Rabbit anti-Human, Mouse, Rat, Clone: 10O4V2, Novus Biologicals™
SDP

Catalog No. NB167635 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167635 100 μg
NB167636 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167635 Supplier Novus Biologicals Supplier No. NBP316272100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Kaiso Monoclonal antibody specifically detects Kaiso in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation

Specifications

Antigen Kaiso
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 10O4V2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias KAISOkaiso transcription factor, transcriptional regulator Kaiso, WUGSC:H_DJ525N14.1, zinc finger and BTB domain containing 33, Zinc finger and BTB domain-containing protein 33, ZNF348kaiso, ZNF-kaiso
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 573-672 of human Kaiso (Q86T24).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10009
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.