Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 1 Antibody, DyLight 755, Novus Biologicals™
SDP

Catalog No. NB059677 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB059677 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB059677 Supplier Novus Biologicals Supplier No. NBP299289IR

Rabbit Polyclonal Antibody

Kallikrein 1 Polyclonal specifically detects Kallikrein 1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Kallikrein 1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.35, glandular kallikrein 1, hK1, kallikrein 1, kallikrein 1, renal/pancreas/salivary, kallikrein serine protease 1, kallikrein-1, Kidney/pancreas/salivary gland kallikrein, Klk6, KLKR, Tissue kallikrein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Kallikrein 1 (NP_002248.1; Met1-Ser262).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3816
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.