Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 11 Antibody (121), PE, Novus Biologicals™
SDP

Catalog No. NB306769 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306769 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306769 Supplier Novus Biologicals Supplier No. NBP289630PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Kallikrein 11 Monoclonal antibody specifically detects Kallikrein 11 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Kallikrein 11
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 121
Conjugate PE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation PBS
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Hippostasin, hK11, kallikrein 11, kallikrein-related peptidase 11, MGC33060, PRSS20kallikrein-11, Serine protease 20, TLSPhippostasin, Trypsin-like protease
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 11 (Met 1-Asn 250; Accession#: NP_006844.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 11012
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.