Learn More
Description
Specifications
Specifications
| Antigen | Kallikrein 11 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, Hippostasin, hK11, kallikrein 11, kallikrein-related peptidase 11, MGC33060, PRSS20kallikrein-11, Serine protease 20, TLSPhippostasin, Trypsin-like protease |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Kallikrein 11 (NP_006844.1; Met 1-Asn 250). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
