Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 15 Antibody (05), PerCP, Novus Biologicals™
SDP

Catalog No. NB293377 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293377 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293377 Supplier Novus Biologicals Supplier No. NBP326080PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kallikrein 15 Monoclonal antibody specifically detects Kallikrein 15 in Human samples. It is validated for ELISA

Specifications

Antigen Kallikrein 15
Applications ELISA
Classification Monoclonal
Clone 05
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias ACO, ACO protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, HSRNASPH, kallikrein 15, kallikrein-15, kallikrein-like serine protease, kallikrein-related peptidase 15, prostinogen
Host Species Mouse
Immunogen Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Kallikrein 15 (Accession#: NP_059979.2, Met1-Asn256)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 55554
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.