Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 2 Antibody (SR1547), Novus Biologicals™
SDP

Catalog No. NB258354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258354 50 μg
NB259063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258354 Supplier Novus Biologicals Supplier No. NBP32226050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Kallikrein 2 Monoclonal antibody specifically detects Kallikrein 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Kallikrein 2
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone SR1547
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2
Host Species Rabbit
Immunogen A synthesized peptide derived from human Kallikrein 2 (Uniprot #: P20151)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3817
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.