Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 3/PSA Antibody (A67-B/E3), Novus Biologicals™
SDP

Catalog No. NBP232992F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232992F 0.02 mg
NBP232992A 0.1 mg
NBP232992B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP232992F Supplier Novus Biologicals Supplier No. NBP2329920.02MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kallikrein 3/PSA Monoclonal specifically detects Kallikrein 3/PSA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Kallikrein 3/PSA
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A67-B/E3
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml, SDS-Page
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. P07288
Gene Alias APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin
Gene Symbols KLK3
Host Species Mouse
Immunogen PSA (p30) from human sperm plasma
Purification Method Protein A purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 354
Test Specificity Recognizes a single protein of 33-34kDa, identified as the prostate specific antigen (PSA). This MAb is highly specific to PSA and stains prostatic secretory and ductal epithelium in both normal and neoplastic tissues. PSA is a chymotrypsin-like serine protease (kallikrein family) exclusively produced by the prostate epithelium, and abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia. This MAb makes an excellent pair with MAb 1A7G6B6 for PSA tests.
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.