Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Kallikrein 3/PSA Antibody (SPM352) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB397112 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397112 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397112 Supplier Novus Biologicals Supplier No. NBP2347680.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kallikrein 3/PSA Monoclonal antibody specifically detects Kallikrein 3/PSA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF

Specifications

Antigen Kallikrein 3/PSA
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone SPM352
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1 μg/mL, Immunohistochemistry-Paraffin 0.5-1 μg/mL, CyTOF-ready
Formulation 10 mM PBS
Gene Alias APS seminin, EC 3.4.21, EC 3.4.21.77, Gamma-seminoprotein, hK3, kallikrein 3, (prostate specific antigen), Kallikrein-3, kallikrein-related peptidase 3, KLK2A1, P-30 antigen, prostate specific antigen, prostate-specific antigen, PSA, PSA semenogelase, Semenogelase, Seminin
Host Species Mouse
Immunogen Kallikrein 3/PSA from human sperm plasma (Uniprot: P07288)
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 354
Target Species Human
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.