Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kallikrein 4/Prostase/EMSP1 Antibody (104), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB334919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334919 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334919 Supplier Novus Biologicals Supplier No. NBP290008JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Kallikrein 4/Prostase/EMSP1 Monoclonal antibody specifically detects Kallikrein 4/Prostase/EMSP1 in Human samples. It is validated for ELISA

Specifications

Antigen Kallikrein 4/Prostase/EMSP1
Applications ELISA
Classification Monoclonal
Clone 104
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AI2A1, ARM1, EC 3.4.21, EC 3.4.21.-, EMSP, EMSP1MGC116827, Enamel matrix serine proteinase 1, kallikrein 4 (prostase, enamel matrix, prostate), kallikrein-4, Kallikrein-like protein 1, kallikrein-related peptidase 4, KLK-L1MGC116828, Prostase, PRSS17enamel matrix serine protease 1, PSTSandrogen-regulated message 1, Serine protease 17
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human Kallikrein 4/Prostase/EMSP1 (Accession#: NP_004908.3, Met1-Ser254)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9622
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.